Learn More
N1,O2'-Dimethyladenosine, TRC
Product Code. 30012717
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012717 1 mg 1mg
30004841 5 mg 5mg
30012709 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012717 Supplier Toronto Research Chemicals Supplier No. TRCD4474101MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91101-00-7
Molecular Formula C12H17N5O4
Molecular Weight (g/mol) 295.29
Synonym 1-Methyl-2'-O-methyladenosine, 2'-O-Methyl-1-methyladenosine;
SMILES O[C@H]1[C@@H](OC)[C@H](N2C(N=CN(C)C3=N)=C3N=C2)O[C@@H]1CO
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C12H19N5O4/c1-16-4-15-11-7(10(16)13)14-5-17(11)12-9(20-2)8(19)6(3-18)21-12/h5-6,8-9,12-13,15,18-19H,3-4H2,1-2H3/t6-,8-,9-,12-/m1/s1
Formula Weight 295.1281
Chemical Name or Material N1,O2'-Dimethyladenosine