Learn More
N1,N3-Bis(2,3-dihydroxypropyl)-5-(2-hydroxy-N-(2-hydroxyethyl)acetamido)-2,4,6-triiodoisophthalamide, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 87771-40-2 |
| Molecular Formula | C18H24I3N3O9 |
| MDL Number | MFCD00867959 |
| Quantity | 5 g |
| SMILES | O=C(NCC(O)CO)C1=C(I)C(N(CCO)C(CO)=O)=C(I)C(C(NCC(O)CO)=O)=C1I |
| Molecular Weight (g/mol) | 807.11 |
| Percent Purity | 98% |
| Chemical Name or Material | N1,N3-Bis(2,3-dihydroxypropyl)-5-(2-hydroxy-N-(2-hydroxyethyl)acetamido)-2,4,6-triiodoisophthalamide |