Learn More
n-Undecyl-1,1,2,2-d4 Alcohol, TRC
Specifications
Specifications
| Chemical Name or Material | n-Undecyl-1,1,2,2-d4 Alcohol |
| CAS | 1219803-47-0 |
| Molecular Formula | C11 D4 H20 O |
| Quantity | 5 mg |
| Synonym | 1-Undecan-1,1,2,2-d4-ol, Undecanol-1,1,2,2-d4, n-Undecyl-1,1,2,2-d4 alcohol, 1-Undecanol-1,1,2,2-d4, 1-Undecanol-1,1,2,2-D4, 1-Hydroxyundecane-1,1,2,2-d4 |
| InChI Formula | InChI=1S/C11H24O/c1-2-3-4-5-6-7-8-9-10-11-12/h12H,2-11H2,1H3/i10D2,11D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])CCCCCCCCC |
| IUPAC Name | 1,1,2,2-tetradeuterioundecan-1-ol |
| Molecular Weight (g/mol) | 176.332 |
| Formula Weight | 176.208 |