missing translation for 'onlineSavingsMsg'
Learn More

N-Trityl-deshydroxymethyl Losartan, TRC

Product Code. 30456510
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456510 100 mg 100mg
30454625 1 g 1g
30453991 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456510 Supplier Toronto Research Chemicals Supplier No. TRCT887825100MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797133-13-1
Molecular Formula C40H35ClN6
Molecular Weight (g/mol) 635.21
Synonym 5-[4'-[(2-Butyl-4-chloro-1H-imidazol-1-yl)methyl][1,1'-biphenyl]-2-yl]-2-(triphenylmethyl)-2H-tetrazole
IUPAC Name 5-[2-[4-[(2-butyl-4-chloroimidazol-1-yl)methyl]phenyl]phenyl]-2-trityltetrazole
SMILES CCCCc1nc(Cl)cn1Cc2ccc(cc2)c3ccccc3c4nnn(n4)C(c5ccccc5)(c6ccccc6)c7ccccc7

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C40H35ClN6/c1-2-3-23-38-42-37(41)29-46(38)28-30-24-26-31(27-25-30)35-21-13-14-22-36(35)39-43-45-47(44-39)40(32-15-7-4-8-16-32,33-17-9-5-10-18-33)34-19-11-6-12-20-34/h4-22,24-27,29H,2-3,23,28H2,1H3
Formula Weight 634.2612
Chemical Name or Material N-Trityl-deshydroxymethyl Losartan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.