missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic Acid, Millipore™
Description
Buffer substance TES
Specifications
Specifications
| CAS | 7365-44-8 |
| Melting Point | 226°C to 228°C |
| Quantity | 250 g |
| Solution Type | Buffer |
| MDL Number | MFCD00007532 |
| Synonym | N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid, TES, N-[Tris(hydroxymethyl)-methyl]-2-aminoethanesulfonic acid |
| Molecular Weight (g/mol) | 229.24 |
| Grade | Research |
| Chemical Name or Material | N-[Tris(Hydroxymethyl)-Methyl]-2-Aminoethanesulfonic Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?