missing translation for 'onlineSavingsMsg'
Learn More

N-Trimethylsilylimidazole, 99%, MP Biomedicals™

Product Code. 11448351 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448351 100 mL 100mL
11438351 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448351 Supplier MP Biomedicals Supplier No. 0215217280
  • ONLINE EXCLUSIVE PRICE
    212.00 EUR
    201.60 EUR / 100mL
    Save 10.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
Synonym n-trimethylsilyl imidazole, 1-trimethylsilyl imidazole, 1-trimethylsilyl-1h-imidazole, tsim, 1h-imidazole, 1-trimethylsilyl, n-trimethylsilylimidazole, trimethylsilyl imidazole, 1-trimethylsilylimidazole, n-trimethylsilyl imidazol, imidazole, 1-trimethylsilyl
PubChem CID 28925
ChEBI CHEBI:85063
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

Color Colorless
Density 0.956 g/mL
Assay Percent Range 0.99
Quantity 100 mL
Formula Weight 140.3
Physical Form Liquid
Chemical Name or Material N-Trimethylsilylimidazole

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.