Learn More
N-Trifluoroacetylpropargylamine, TRC
Product Code. 30454992
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454992 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30454992 Supplier Toronto Research Chemicals Supplier No. TRCT7896301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Trifluoroacetylpropargylamine
CAS 14719-21-2
Molecular Formula C5H4F3NO
Quantity 1 g
Synonym Trifluoro-N-prop-2-ynylacetamide, 2,2,2-Trifluoro-N-(prop-2-ynyl)acetamide, 2,2,2-Trifluoro-N-prop-2-yne-1-ylacetamide, N-Propargyltrifluoroacetamide, N-Propargyltrifluoroacetylamide, 2,2,2-Trifluoro-N-2-propynylacetamide, 2,2,2-Trifluoro-N-2-propyn-1-ylacetamide
InChI Formula InChI=1S/C5H4F3NO/c1-2-3-9-4(10)5(6,7)8/h1H,3H2,(H,9,10)
SMILES FC(F)(F)C(=O)NCC#C
IUPAC Name 2,2,2-trifluoro-N-prop-2-ynylacetamide
Molecular Weight (g/mol) 151.09
Formula Weight 151.0245