Learn More
N-Trifluoroacetyliso Deacetylcolchicine, TRC
Product Code. 30456556
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456556 Supplier Toronto Research Chemicals Supplier No. TRCT788100100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Trifluoroacetyliso Deacetylcolchicine
CAS 71324-48-6
Molecular Formula C22 H22 F3 N O6
Quantity 100 mg
InChI Formula InChI=1S/C22H22F3NO6/c1-29-16-10-13-12(6-8-15(16)27)18-11(5-7-14(13)26-21(28)22(23,24)25)9-17(30-2)19(31-3)20(18)32-4/h6,8-10,14H,5,7H2,1-4H3,(H,26,28)/t14-/m0/s1
SMILES COC1=CC2=C(C=CC1=O)c3c(CC[C@@H]2NC(=O)C(F)(F)F)cc(OC)c(OC)c3OC
IUPAC Name 2,2,2-trifluoro-N-[(7S)-1,2,3,9-tetramethoxy-10-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Molecular Weight (g/mol) 453.41
Formula Weight 453.1399