Learn More
N-Tricyclo[3.3.1.13,7]dec-1-yl-1H-Indazole-3-carboxamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Tricyclo[3.3.1.13,7]dec-1-yl-1H-Indazole-3-carboxamide |
| CAS | 516445-83-3 |
| Molecular Formula | C18H21N3O |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C18H21N3O/c22-17(16-14-3-1-2-4-15(14)20-21-16)19-18-8-11-5-12(9-18)7-13(6-11)10-18/h1-4,11-13H,5-10H2,(H,19,22)(H,20,21)/t11-,12+,13-,18? |
| SMILES | O=C(NC12C[C@@H]3C[C@@H](C[C@@H](C3)C1)C2)c4n[nH]c5ccccc45 |
| IUPAC Name | N-(1-adamantyl)-1H-indazole-3-carboxamide |
| Molecular Weight (g/mol) | 295.38 |
| Formula Weight | 295.1685 |