missing translation for 'onlineSavingsMsg'
Learn More

n-Propylurea, 98%

Product Code. p-7080578
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418720 5 g 5g
11409442 25 g 25g
11419442 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418720 Supplier Thermo Scientific Alfa Aesar Supplier No. A10160.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 627-06-5 | C4H10N2O | 102.137 g/mol

n-Propylurea is used as a reactant in the preparation of N-1 monosubstituted 8-pyrazolyl xanthines as high affinity A2B adenosine receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propylurea is used as a reactant in the preparation of N-1 monosubstituted 8-pyrazolyl xanthines as high affinity A2B adenosine receptor antagonists.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-06-5
Molecular Formula C4H10N2O
Molecular Weight (g/mol) 102.137
MDL Number MFCD00014793
InChI Key ZQZJKHIIQFPZCS-UHFFFAOYSA-N
Synonym n-propylurea, 1-propylurea, urea, propyl, urea, n-propyl, propyl urea, unii-0pk51j4av7, n-propyl urea, n'-propylurea, amino-n-propylamide, acmc-1ato6
PubChem CID 12303
IUPAC Name propylurea
SMILES CCCNC(=O)N

Specifications

Melting Point 99°C to 104°C
Linear Formula CH3CH2CH2NHCONH2
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 102.14
Percent Purity 98%
Chemical Name or Material n-Propylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.