missing translation for 'onlineSavingsMsg'
Learn More

n-Phenylanthranilic Acid, Sigma-Aldrich™

Product Code. 15821311
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15821311 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 15821311 Supplier Sigma Aldrich Supplier No. 8209790100
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 91-40-7
Melting Point 182°C to 185°C
Molecular Formula 2-(C6H5NH)C6H4COOH
MDL Number MFCD00002421
Quantity 100 g
Synonym N-Phenylanthranilic acid, Diphenylamine-2-carboxylic acid, N-Phenylanthranilic acid
Molecular Weight (g/mol) 213.23
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.