Learn More
N-(p-Toluenesulfonyl)urea, TRC
Specifications
Specifications
| Chemical Name or Material | N-(p-Toluenesulfonyl)urea |
| CAS | 1694-06-0 |
| Molecular Formula | C8 H10 N2 O3 S |
| Quantity | 50 g |
| Synonym | N-(p-Tolylsulfonyl)urea, Tosylurea, (p-Toluenesulfonyl)urea, (p-Tolylsulfonyl)urea, H 16650, (p-Tolylsulfonyl)urea, N-(Aminocarbonyl)-4-methylbenzenesulfonamide |
| InChI Formula | InChI=1S/C8H10N2O3S/c1-6-2-4-7(5-3-6)14(12,13)10-8(9)11/h2-5H,1H3,(H3,9,10,11) |
| SMILES | Cc1ccc(cc1)S(=O)(=O)NC(=O)N |
| IUPAC Name | p-tolylsulfonylurea |
| Molecular Weight (g/mol) | 214.242 |
| Formula Weight | 214.0412 |
Product Title
Haben Sie Verbesserungsvorschläge?