Learn More
n-Octane, Sigma-Aldrich™
Product Code. 15849360
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849360 100 mL 100mL
15806430 250 mL 250mL
15816430 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.
We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15849360 Supplier Sigma Aldrich Supplier No. 8069100100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Dieses Produkt unterliegt der Verbrauchsteuer auf Energieerzeugnisse gemäß der europäischen Richtlinie 2003/96 / EWG.
For synthesis
n-octane is a hydrocarbon compound used as a solvent, cleaning agent, fuel additive, or reaction agents.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 111-65-9
Spezifikation
Melting Point -57°C
Density 0.70 g/cm3 at 20°C
Boiling Point 125°C to 126°C/1013 hPa
Molecular Formula CH3(CH2)6CH3
MDL Number MFCD00009556
Quantity 100 mL
Synonym n-Octane, n-Octane
Molecular Weight (g/mol) 114.23
Grade Research