missing translation for 'onlineSavingsMsg'
Learn More

N,O-Dimethylhydroxylamine Hydrochloride, Sigma-Aldrich™

Product Code. 15861861
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861861 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861861 Supplier Sigma Aldrich Supplier No. 8400740010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 6638-79-5
Melting Point 110°C to 116°C
MDL Number MFCD00012485
Quantity 10 g
Synonym N,O-Dimethylhydroxylamine hydrochloride, N-Methoxymethanamine hydrochloride, N,O-Dimethylhydroxylamine hydrochloride
Molecular Weight (g/mol) 61.08 (Free Base Basis)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.