missing translation for 'onlineSavingsMsg'
Learn More

N-O-bis-(Trimethylsilyl)trifluoroacetamide, 99.26%, MP Biomedicals™

Product Code. 11487721 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487721 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487721 Supplier MP Biomedicals Supplier No. 0215047605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N-O-bis-(Trimethylsilyl)trifluoroacetamide is a silylating agent for amino acids, carbohydrates, and steroids.

  • N-O-bis-(Trimethylsilyl)trifluoroacetamide is a derivatization reagent employed in a GC-MS analysis of furfural content in foods.
TRUSTED_SUSTAINABILITY

Specifications

CAS 21149-38-2
Color Colorless
Molecular Formula C8H18F3NOSi2
MDL Number MFCD00008269
Quantity 5 g
Synonym bis trimethylsilyl trifluoroacetamide, 2,2,2-trifluoro-n,n-bis trimethylsilyl acetamide, acetamide, 2,2,2-trifluoro-n,n-bis trimethylsilyl, unii-5t8na426kz, n,n-bstfa, ksc497c2d, bistrimethylsilyltrifluoroacetamide, bis-trimethylsilyltrifluoroacetamide, bistrimethylsilyl-trifluoroacetamide
InChI Key RZYHXKLKJRGJGP-UHFFFAOYSA-N
SMILES C[Si](C)(C)N(C(=O)C(F)(F)F)[Si](C)(C)C
IUPAC Name 2,2,2-trifluoro-N,N-bis(trimethylsilyl)acetamide
Molecular Weight (g/mol) 257.403
PubChem CID 65279
Formula Weight 257.4
Physical Form Liquid
Chemical Name or Material N-O-bis-(Trimethylsilyl)trifluoroacetamide, FTIR
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.