Learn More
N,O-Bis(trimethylsilyl)acetamide, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | N,O-Bis(trimethylsilyl)acetamide |
| CAS | 10416-59-8 |
| Molecular Formula | C8H21NOSi2 |
| Quantity | 100 g |
| Synonym | BSA, BSiA, Bis(trimethylsilyl)acetamide, Bis(trimethylsilyl)acetimidate, Dynasylan, BSA, LS 7270, N,O-Bis(trimethylsillyl)acetamide, N-(Trimethylsilyl)acetimidic Acid Trimethylsilyl Ester, N-(Trimethylsilyl)ethanimidic Acid Trimethylsilyl Ester, O,N-Bis(trimethylsilyl)acetamide |
| InChI Formula | InChI=1S/C8H21NOSi2/c1-8(9-11(2,3)4)10-12(5,6)7/h1-7H3 |
| SMILES | C\C(=N/[Si](C)(C)C)\O[Si](C)(C)C |
| IUPAC Name | trimethylsilyl N-trimethylsilylethanimidate |
| Molecular Weight (g/mol) | 203.43 |
| Formula Weight | 203.1162 |