Learn More
N-(Nitrosomethyl)ethanolamine, TRC
Product Code. 30004170
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004170 250 mg 250mg
30010000 50 mg 50mg
18071530 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004170 Supplier Toronto Research Chemicals Supplier No. TRCN589055250MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26921-68-6
Molecular Formula C3H8N2O2
Molecular Weight (g/mol) 104.11
Synonym Ethanol, 2-(methylnitrosamino)- (6CI,7CI,8CI), 2-(Methylnitrosoamino)ethanol, Methyl(2-hydroxyethyl)nitrosamine, Methyl(2-hydroxyethyl)nitrosoamine, N-(Nitrosomethyl)ethanolamine, N-Methyl-N-nitrosoethanolamine, N-Nitrosomethyl(2-hydroxyethyl)amine
IUPAC Name N-(2-hydroxyethyl)-N-methylnitrous amide
SMILES CN(CCO)N=O
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C3H8N2O2/c1-5(4-7)2-3-6/h6H,2-3H2,1H3
Formula Weight 104.0586
Percent Purity >95%
Chemical Name or Material N-Nitroso-N-methylethanolamine
Grade HPLC