Learn More
N-Nitroso Moexipril, TRC
Product Code. 30415372
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30415372 100 mg 100mg
30415063 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30415372 Supplier Toronto Research Chemicals Supplier No. TRCQ825865100MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Nitroso Moexipril
CAS 1026879-47-9
Molecular Formula C27H33N3O8
Quantity 100 mg
Synonym (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]nitrosoamino]-1-oxopropyl]-3,4-dihydro-6,7-dimethoxy-3(1H)-isoquinolinecarboxylic acid, (S)-2-(N-((S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
InChI Formula InChI=1S/C27H33N3O8/c1-5-38-27(34)21(12-11-18-9-7-6-8-10-18)30(28-35)17(2)25(31)29-16-20-15-24(37-4)23(36-3)14-19(20)13-22(29)26(32)33/h6-10,14-15,17,21-22H,5,11-13,16H2,1-4H3,(H,32,33)/t17-,21-,22-/m0/s1
SMILES C([C@@H](N([C@@H](CCC1=CC=CC=C1)C(OCC)=O)N=O)C)(=O)N2CC=3C(C[C@H]2C(O)=O)=CC(OC)=C(OC)C3
IUPAC Name (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]nitrosoamino]-1-oxopropyl]-3,4-dihydro-6,7-dimethoxy-3(1H)-isoquinolinecarboxylic acid
Molecular Weight (g/mol) 527.57
Formula Weight 527.23