Learn More
N-Nitroso Delapril, TRC
Specifications
Specifications
| Chemical Name or Material | N-Nitroso Delapril |
| CAS | 1027955-20-9 |
| Molecular Formula | C26 H31 N3 O6 |
| Quantity | 100 mg |
| Synonym | N-(2,3-Dihydro-1H-inden-2-yl)-N-(N-((S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl)-N-nitroso-L-alanyl)glycine |
| InChI Formula | InChI=1S/C26H31N3O6/c1-3-35-26(33)23(14-13-19-9-5-4-6-10-19)29(27-34)18(2)25(32)28(17-24(30)31)22-15-20-11-7-8-12-21(20)16-22/h4-12,18,22-23H,3,13-17H2,1-2H3,(H,30,31)/t18-,23-/m0/s1 |
| SMILES | CCOC(=O)[C@H](CCc1ccccc1)N(N=O)[C@@H](C)C(=O)N(CC(=O)O)C2Cc3ccccc3C2 |
| IUPAC Name | 2-[2,3-dihydro-1H-inden-2-yl-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]-nitrosoamino]propanoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 481.541 |
| Formula Weight | 481.221 |