Learn More
N,N,N′,N′-Tetramethyl-1,4-phenylenediamine, Sigma-Aldrich™
Product Code. 15841391
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15841391 25 g 25g
15831391 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15841391 Supplier Sigma Aldrich Supplier No. 8211010025
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100-22-1
Specifications
Melting Point 48°C to 50°C
Molecular Formula C6H4-1,4-[N(ch3)2]2
MDL Number MFCD00008309
Quantity 25 g
Synonym N,N,N′,N′-Tetramethyl-1,4-phenylenediamine, 1,4-Bis(dimethylamino)benzene, N,N,N′,N′-Tetramethyl-1,4-phenylenediamine
Molecular Weight (g/mol) 164.25
Grade Research