Learn More
N,N'-Dinitrosopenta-methylenetetramine, Pure, MP Biomedicals
Product Code. 11422475 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422475 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11422475 Supplier MP Biomedicals Supplier No. 0520631410
 Restricted Item
Estimated Shipment: 23-12-2026
Specifications
CAS 101-25-7
Molecular Formula C5H10N6O2
Quantity 10 g
Synonym dipentax, micropor, dnpmt, opex, mikrofor n, porophor b, dinitrosopentamethylenetetramine, vulcacel bn, unicel-nd, unicel ndx
InChI Key MWRWFPQBGSZWNV-UHFFFAOYSA-N
SMILES C1N2CN(CN1CN(C2)N=O)N=O
IUPAC Name 3,7-dinitroso-1,3,5,7-tetrazabicyclo[3.3.1]nonane
Molecular Weight (g/mol) 186.175
PubChem CID 7549
ChEBI CHEBI:82457
Formula Weight 186.2
Chemical Name or Material N, N«-Dinitrosopenta-Methylenetetramine, Pure
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N'-DINITROSOPENTA-METHYLENETETRAMINE
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 3,7-dinitroso-1,3,5,7-tetraazabicyclo[3.3.1]nonane