Learn More
N,N-Dimethylethylenediamine, Sigma-Aldrich™
Description
N,N-Dimethylethylenediamine (DMEDA) is used as a bidentate ligand in coordination chemistry due to the presence of two nitrogen atoms, one with lone pairs that can coordinate to a metal center. It is used as a ligand and base in Suzuki-Miyaura and Sonogashira coupling reactions, enhancing palladium catalyst stability, and deprotonating alkynes. It acts as a nucleophile and a base, promoting reactions involving proton transfer or nucleophilic attack. It is commonly used in amination reactions to synthesize substituted amines and in epoxide ring openings to form amino alcohols. Additionally, DMEDA serves as a chelating agent, forming stable complexes with transition metals. It acts as a secondary amine in reductive amination to synthesize tertiary amines.
Specifications
Specifications
| CAS | 108-00-9 |
| Melting Point | <-70°C |
| Density | 0.81 g/cm3 at 20°C |
| Boiling Point | 107°C to 109°C/1013 hPa |
| Molecular Formula | NH2C2H4N(CH3)2 |
| MDL Number | MFCD00008175 |
| Quantity | 100 mL |
| Synonym | N,N-Dimethylethylenediamine, 2-(Dimethylamino)ethylamine, 1-Amino-2-dimethylaminoethane, N,N-Dimethylethylenediamine |
| Molecular Weight (g/mol) | 88.15 |
| Grade | Research |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.