missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylethylamine, 99%

Product Code. p-7027324
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456886 100 mL 100mL
11466886 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456886 Supplier Thermo Scientific Alfa Aesar Supplier No. B23992.AE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-56-1 | C4H11N | 73.139 g/mol

N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylethylamine is used as adhesive, sealant chemical and process regulator. It acts as an intermediate for quaternary ammonium compounds and is also used as a stabilizer for chlorinated hydrocarbons and vinyl derivatives. It is used in foundries as a catalyst for sand core production. Further, it is employed in the polymerization of polyamides. In addition to this, it is used in the preparation of polypeptide nanogels with self-reinforced endocytoses.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents and peroxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-56-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009039
InChI Key DAZXVJBJRMWXJP-UHFFFAOYSA-N
Synonym n,n-dimethylethylamine, dimethylethylamine, ethanamine, n,n-dimethyl, ethyldimethylamine, n-ethyldimethylamine, ethylamine, n,n-dimethyl, methanamine, n-ethyl-n-methyl, unii-9n5384xvem, c13-15-alkyldimethylamines, amines, c13-15-alkyldimethyl
PubChem CID 11723
IUPAC Name N,N-dimethylethanamine
SMILES CCN(C)C

Specifications

Melting Point -140°C
Density 0.675
Boiling Point 36°C to 37°C
Flash Point −36°C (−33°F)
Odor Ammonia-like
Linear Formula CH3CH2N(CH3)2
Refractive Index 1.373
Quantity 100 mL
UN Number UN2733
Beilstein 1696893
Sensitivity Air sensitive
Solubility Information Miscible with water.
Formula Weight 73.14
Percent Purity 99%
Chemical Name or Material N,N-Dimethylethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Dimethyl ethylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.