Learn More
N,N-Dimethylallylamine, Sigma-Aldrich™
Specifications
Specifications
| CAS | 2155-94-4 |
| Density | 0.730 g/cm3 at 20°C |
| Molecular Formula | CH2=CHCH2N(CH3)2 |
| MDL Number | MFCD00038234 |
| Quantity | 25 mL |
| Synonym | N,N-Dimethylallylamine, N-Allyldimethylamine, N,N-Dimethylallylamine |
| Molecular Weight (g/mol) | 85.15 |
| Grade | Research |