missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, Sigma-Aldrich™

Product Code. 15896350
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896350 2.5 L 2.5 L
15886350 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896350 Supplier Sigma Aldrich Supplier No. 8032352500
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

N,N-Dimethylacetamide (DMAc) is a water-miscible organic solvent used in the synthesis of styrene, vinyl resin, linear polyester, acrylic fiber, plastic, pesticide, and synthetic leather. It participates in dehydrogenation, dehydration, halogenation, deprotonation reactions and reductive carbonylation of transition metals. It can also act as catalysts, promoters, halogen/alkoxy or hydroxyl/halogen exchange mediators, reducing agents, and sources of the base.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-19-5

Specifications

Melting Point -20°C
Density 0.94 g/cm3 at 20°C
Quantity 2.5 L
Molecular Formula CH3CON(CH3)2
MDL Number MFCD00008686
Synonym N,N-Dimethylacetamide, Acetic acid dimethylamide, Acetyldimethylamine, DMA, N,N-Dimethylacetamide
Boiling Point 165°C to 166°C/1013 hPa
Molecular Weight (g/mol) 87.12
Grade Research
Chemical Name or Material N,N-Dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.