missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, MilliporeSigma™

Product Code. 15823880
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15823880 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15823880 Supplier Supelco Supplier No. 1003991000
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For headspace gas chromatography SupraSolv™

TRUSTED_SUSTAINABILITY

Specifications

pH 4 (at 200g/L 20°C)
Melting Point -20°C
CAS 127-19-5
Color Colorless
Density 0.94g/cm3 at 20°C
Quantity 1 L
Flash Point 64°C (147.2°F)
Ignition Point 400°C (752°F)
Odor Amine-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.423
Molecular Formula C4H9NO
Vapor Pressure 1.76 hPa (20°C)
MDL Number MFCD00008686
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
Boiling Point 165°C to 166°C
Solubility Information Soluble in water at 20°C
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILES CN(C)C(C)=O
IUPAC Name N,N-dimethylacetamide
Molecular Weight (g/mol) 87.12
PubChem CID 31374
ChEBI CHEBI:84254
Formula Weight 87.12
Chemical Name or Material N,N-Dimethylacetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.