missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylnicotinamide, MP Biomedicals

Product Code. 11433045 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433045 100 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11433045 Supplier MP Biomedicals Supplier No. 0521149825

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 59-26-7
Molecular Formula C10H14N2O
Quantity 100 g
Synonym nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
InChI Key NCYVXEGFNDZQCU-UHFFFAOYSA-N
SMILES CCN(CC)C(=O)C1=CN=CC=C1
IUPAC Name N,N-diethylpyridine-3-carboxamide
Molecular Weight (g/mol) 178.235
PubChem CID 5497
Formula Weight 178.24
Chemical Name or Material N, N-Diethylnicotinamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.