Learn More
N,N-Diethyl-1,4-butanediamine, 96%
Product Code. 11479410
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11479410 1 g 1g
11489410 5 g 5g
2 options
Product Code. 11479410
Brand: Thermo Scientific Alfa Aesar H50278.03
Request Bulk or Custom Format
CAS: 27431-62-5 | C8H20N2 | 144.262 g/mol
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27431-62-5
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00014836
InChI Key JILXUIANNUALRZ-UHFFFAOYSA-N
Synonym 4-aminobutyl diethylamine, 4-diethylaminobutylamine, 4-diethylamino butylamine, n1,n1-diethylbutane-1,4-diamine, n,n-diethylbutane-1,4-diamine, n,n-diethyl-1,4-butanediamine, n 1 ,n 4-diethylputrescine, 1,4-butanediamine, n,n-diethyl, diethylaminobutylamine, acmc-1cneh
PubChem CID 117976
IUPAC Name N',N'-diethylbutane-1,4-diamine
SMILES CCN(CC)CCCCN
Specifications
Density 0.83
Boiling Point 72°C to 24°C (10 mmHg)
Linear Formula (CH3CH2)2NCH2CH2CH2CH2NH2
Quantity 1 g
UN Number UN2735
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 144.26
Percent Purity 96%
Chemical Name or Material N,N-Diethyl-1,4-butanediamine