Learn More
(+)-N,N'-Dibenzyl-L-tartaric diamide, 99%, 99% e.e., Thermo Scientific™
Product Code. 10471484 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10471484 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10471484 Supplier Thermo Scientific Acros Supplier No. 426980050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 88393-56-0
CAS Min % 98.5
CAS Max % 100.0
Melting Point 202°C to 206°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C18H20N2O4
MDL Number MFCD00064478
Quantity 5 g
Specific Rotation +80° to +86° (20°C, 589nm) (c=1, DMF)
Synonym 2r,3r-n1,n4-dibenzyl-2,3-dihydroxysuccinamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxysuccinamide, dibenzyl tartramide, n,n'-dibenzyl-l-tartramide, +-n,n-dibenzyl-l-tartaric diamide, +-n,n'-dibenzyl-l-tartaric diamide, 2r,3r-n,n'-dibenzyl-2,3-dihydroxybutanediamide, n,n inverted exclamation marka-dibenzyl-l-tartramide, +-n,n inverted exclamation marka-dibenzyl-l-tartaric diamide
InChI Key BBYSAHVLSFBCMN-MNSHXRSINA-N
SMILES O[C@H]([C@@H](O)C(=O)NCC1=CC=CC=C1)C(=O)NCC1=CC=CC=C1
IUPAC Name (2R,3R)-N,N'-dibenzyl-2,3-dihydroxybutanediamide
Molecular Weight (g/mol) 328.37
PubChem CID 7271811
Formula Weight 328.37
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material (+)-N, N'-Dibenzyl-L-tartaric diamide, 99% ee