Learn More
N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 42137-88-2 |
| Molecular Formula | C11H15Cl2NO2S |
| MDL Number | MFCD00018944 |
| Quantity | 10 g |
| SMILES | O=S(C1=CC=C(C)C=C1)(N(CCCl)CCCl)=O |
| Molecular Weight (g/mol) | 296.21 |
| Percent Purity | 95% |
| Chemical Name or Material | N,N-Bis(2-chloroethyl)-4-methylbenzenesulfonamide |