Learn More
N-Methylethylenediamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methylethylenediamine |
| CAS | 109-81-9 |
| Molecular Formula | C3H10N2 |
| Quantity | 100 mg |
| Synonym | N1-Methyl-1,2-ethanediamine, (2-Aminoethyl)methylamine, 1-(Methylamino)-2-aminoethane, 1-Methylethylenediamine, 2-(Methylamino)ethanamine, 2-(Methylamino)ethylamine, N-Methyl-1,2-diaminoethane, N-Methyl-1,2-ethanediamine, N-Methyl-1,2-ethylenediamine |
| InChI Formula | InChI=1S/C3H10N2/c1-5-3-2-4/h5H,2-4H2,1H3 |
| SMILES | CNCCN |
| IUPAC Name | N'-methylethane-1,2-diamine |
| Molecular Weight (g/mol) | 74.12 |
| Formula Weight | 74.0844 |