missing translation for 'onlineSavingsMsg'
Learn More

N-Methylbenzenesulfonamide, 97%

Product Code. 15471978
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471978 250 mg 250mg
15461978 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471978 Supplier Thermo Scientific Alfa Aesar Supplier No. H55666.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5183-78-8 | C7H9NO2S | 171.21 g/mol

N-Methylbenzenesulfonamide, is used as an intermediate in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylbenzenesulfonamide, is used as an intermediate in industry.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5183-78-8
Molecular Formula C7H9NO2S
Molecular Weight (g/mol) 171.21
MDL Number MFCD00025023
InChI Key SVDVKEBISAOWJT-UHFFFAOYSA-N
Synonym benzenesulfonamide, n-methyl, n-methylbenzenesulphonamide, n-phenylsulfonyl methanamine, methyl phenylsulfonyl amine, n-methylphenylsulfonamide, acmc-1avn9, n-methylbenzene sulfonamide, n-methyl benzenesulphonamide, benzenesulfonamide,n-methyl, benzenesulfonyl-n-methylamide
PubChem CID 78858
IUPAC Name N-methylbenzenesulfonamide
SMILES CNS(=O)(=O)C1=CC=CC=C1

Specifications

Quantity 250 mg
Solubility Information It is soluble in water.
Formula Weight 171.21
Percent Purity 97%
Chemical Name or Material N-Methylbenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.