Learn More
N-Methyl-N-trimethylsilylacetamide, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-N-trimethylsilylacetamide |
| CAS | 7449-74-3 |
| Molecular Formula | C6H15NOSi |
| Quantity | 1 g |
| Synonym | N-Trimethylsilyl-N-methylacetamide, NSC 142435 |
| InChI Formula | InChI=1S/C6H15NOSi/c1-6(8)7(2)9(3,4)5/h1-5H3 |
| SMILES | CN(C(=O)C)[Si](C)(C)C |
| IUPAC Name | N-methyl-N-trimethylsilylacetamide |
| Molecular Weight (g/mol) | 145.27 |
| Formula Weight | 145.0923 |