Learn More
N-Methyl-N-(quinolin-6-ylmethyl)amine, 95%, Thermo Scientific™
Product Code. 10366844
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10366844 250 mg 250mg
10778045 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10366844 Supplier Thermo Scientific Maybridge Supplier No. CC04546CB
 Restricted Item
Estimated Shipment: 05-01-2027
Chemical Identifiers
CAS 179873-36-0
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.23
MDL Number MFCD06738897
InChI Key IIPNTNDPIZNFRU-UHFFFAOYSA-N
Synonym n-methyl-n-quinolin-6-ylmethyl amine, n-methyl-1-quinolin-6-yl methanamine, methyl quinolin-6-ylmethyl amine, methyl-quinolin-6-ylmethyl-amine, n-methyl-n-6-quinolinylmethyl amine, 6-quinolinemethanamine, n-methyl, 6-quinolinemethanamine,n-methyl, n-methyl-1-6-quinolinyl methanamine, n-methyl-6-quinolinemethanamine, methyl 6-quinolylmethyl amine
PubChem CID 6483903
IUPAC Name N-methyl-1-quinolin-6-ylmethanamine
SMILES CNCC1=CC=C2N=CC=CC2=C1
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(quinolin-6-ylmethyl)amine