Learn More
N-Methyl-N-(o-aminophenyl)anthranilic Acid Methyl Ester, TRC
Product Code. 30475708
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475708 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30475708 Supplier Toronto Research Chemicals Supplier No. TRCM286955100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-N-(o-aminophenyl)anthranilic Acid Methyl Ester
CAS 1346601-40-8
Molecular Formula C15H16N2O2
Quantity 100 mg
Synonym 2-Methyl-2-(o-aminophenyl)amino Benzoic Acid Methyl Ester
InChI Formula InChI=1S/C15H16N2O2/c1-17(14-10-6-4-8-12(14)16)13-9-5-3-7-11(13)15(18)19-2/h3-10H,16H2,1-2H3
SMILES COC(=O)c1ccccc1N(C)c2ccccc2N
IUPAC Name methyl 2-(2-amino-N-methylanilino)benzoate
Molecular Weight (g/mol) 256.3
Formula Weight 256.1212