missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-L-glutamic acid, MP Biomedicals™

Product Code. 11434781 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434781 1 g 1g
11424781 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11434781 Supplier MP Biomedicals Supplier No. 0215555591
  • ONLINE EXCLUSIVE PRICE
    433.00 EUR
    411.60 EUR / 1g
    Save 21.40 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6753-62-4
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00065920
InChI Key LAYOOVKHWQMRIQ-WCCKRBBISA-N
Synonym n-me-glu-oh, h-l-meglu-oh hcl, n-alpha-methyl-l-glutamic acid hydrochloride, n-methyl-l-glutamic acid hydrochloride, s-2-methylamino pentanedioic acid hydrochloride
PubChem CID 56777325
IUPAC Name (2S)-2-(methylamino)pentanedioic acid;hydrochloride
SMILES CNC(CCC(=O)O)C(=O)O.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 161.16
Physical Form Solid
Chemical Name or Material N-methyl-L-glutamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.