Learn More
N-Methyl β-Hydroxyl Phentermine, TRC
Specifications
Specifications
| CAS | 10079-55-7 |
| Molecular Formula | C11 H17 N O |
| Quantity | 25 mg |
| Synonym | Oxethazaine TLC Impurity 2 |
| InChI Formula | InChI=1S/C11H17NO/c1-11(2,12-3)10(13)9-7-5-4-6-8-9/h4-8,10,12-13H,1-3H3 |
| SMILES | CNC(C)(C)C(O)c1ccccc1 |
| IUPAC Name | 2-methyl-2-(methylamino)-1-phenyl-propan-1-ol |
| Molecular Weight (g/mol) | 179.26 |
| Formula Weight | 179.131 |
| Chemical Name or Material | 2-methyl-2-(methylamino)-1-phenyl-propan-1-ol |