Learn More
N-methyl-d3-homopiperazine, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-1,4-diazepane-D3 (N-methyl-D3) |
| CAS | 2488857-81-2 |
| Molecular Formula | C6 D3 H11 N2 |
| Quantity | 0.05 g |
| Synonym | Hexahydro-1-(methyl-d3)-1 H-1,4-diazepine, 1-(Methyl-d3)-1,4-diazepane, 1-Trideuteriomethyl-2,3,4,5,6,7-hexahydro-1 H-1,4-diazepine, 1-Methyl-d3-homopiperazine, N-(Methyl-d3)-1,4-diazacycloheptane, N-(Methyl-d3)homopiperazine, 1-Methyl-1,4-diazepane-D3 (N-methyl-D3) |
| InChI Formula | InChI=1 S/C6H14N2/c1-8-5-2-3-7-4-6-8/h7H,2-6H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CCCNCC1 |
| IUPAC Name | 1-(trideuteriomethyl)-1,4-diazepane |
| Molecular Weight (g/mol) | 117.2073 |
| Formula Weight | 117.1345 g/mol |
| Show More |