missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-5-chloroisothiazolone (5-Chloro-2-methyl-4-isothiazolin-3-one), Mikromol™

Product Code. 18032010
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18032010 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032010 Supplier Mikromol Supplier No. MM0877.060100
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Methyl-5-chloroisothiazolone (5-Chloro-2-methyl-4-isothiazolin-3-one)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Chloro-2-methyl-4-isothiazolin-3-one
CAS 26172-55-4
Analyte or Component Names 5-Chloro-2-methyl-4-isothiazolin-3-one
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H4 Cl N O S
Certifications/Compliance ISO 17025
Synonym 5-Chloro-2-methyl-4-isothiazolin-3-one, 2-Methyl-5-chloro-3-isothiazolone, 2-Methyl-5-chloroisothiazolin-3-one, 5-Chloro-2-methyl-2H-isothiazol-3-one, 5-Chloro-2-methyl-3(2H)-isothiazolone, 5-Chloro-2-methyl-3-isothiazolone, 5-Chloro-2-methyl-4-isothiazolin-3-one, 5-Chloro-2-methylisothiazolin-3-one, 5-Chloro-N-methylisothiazolin-3-one, 5-Chloro-N-methylisothiazolone, A 33 (bactericide), Bioace, CIT, CMIT, HS 818 (antiseptic), Kathon CG 5243, Kathon IXE, Methylchloroisothiazolinone, N-Methyl-5-chloroisothiazolin-3-one, N-Methyl-5-chloroisothiazolone, T 360
InChI Formula InChI=1S/C4H4ClNOS/c1-6-4(7)2-3(5)8-6/h2H,1H3
SMILES CN1SC(=CC1=O)Cl
IUPAC Name 5-chloro-2-methyl-1,2-thiazol-3-one
Molecular Weight (g/mol) 149.6
Quantity 100 mg
Formula Weight 148.9702 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.