Learn More
N-Methyl-(2-bromo-3-furyl)methylamine hydrochloride, 97%, Thermo Scientific™
Product Code. 10602315
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602315 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10602315 Supplier Thermo Scientific Maybridge Supplier No. CC64796DA
 Restricted Item
Estimated Shipment: 16-12-2026
Specifications
CAS 926921-76-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H9BrClNO
MDL Number MFCD09879945
Quantity 1 g
Synonym 2-bromo-3-methylamino methyl furan hydrochloride, 2-bromofuran-3-yl methyl methyl amine hydrochloride, 2-bromo 3-furyl methyl methylamine, chloride, 1-2-bromofuran-3-yl-n-methylmethanamine hydrochloride, 1-2-bromofuran-3-yl-n-methylmethanamine-hydrogen chloride 1/1
InChI Key GYLBKUAMHLATCB-UHFFFAOYSA-N
SMILES CNCC1=C(OC=C1)Br.Cl
IUPAC Name 1-(2-bromofuran-3-yl)-N-methylmethanamine;hydrochloride
Molecular Weight (g/mol) 226.498
PubChem CID 24229719
Formula Weight 226.5
Percent Purity 97%
Chemical Name or Material N-Methyl-(2-bromo-3-furyl)methylamine hydrochloride