Learn More
N-Maleimidomethanol, TRC
Product Code. 30011861
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011861 100 mg 100mg
30006052 1 g 1g
30005765 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011861 Supplier Toronto Research Chemicals Supplier No. TRCM138675100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5063-96-7
Molecular Formula C5 H5 N O3
Molecular Weight (g/mol) 127.1
IUPAC Name 1-(hydroxymethyl)pyrrole-2,5-dione
SMILES OCN1C(=O)C=CC1=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C5H5NO3/c7-3-6-4(8)1-2-5(6)9/h1-2,7H,3H2
Formula Weight 127.0269
Chemical Name or Material N-Maleimidomethanol