Learn More
N-(Hydroxymethyl)acetamide, TRC
Product Code. 30010816
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010816 500 mg 500mg
30003507 2.5 g 2.5g
30007912 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010816 Supplier Toronto Research Chemicals Supplier No. TRCH973010500MG
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 625-51-4
Molecular Weight (g/mol) 89.09
Synonym Acetamidomethanol, Formicin, Monomethylolacetamide, N-Methylolacetamide, N-Monomethylolacetamide
SMILES CC(NCO)=O
Specifications
Molecular Formula C3H7NO2
Quantity 500 mg
InChI Formula InChI=1S/C3H7NO2/c1-3(6)4-2-5/h5H,2H2,1H3,(H,4,6)
Formula Weight 89.0477
Chemical Name or Material N-(Hydroxymethyl)acetamide