Learn More
n-Heptyl 4-Hydroxybenzoate, TRC
Product Code. 30406320
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406320 100 g -
30393082 5 g 5g
30389922 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30406320 Supplier Toronto Research Chemicals Supplier No. TRCH283180100G
 Restricted Item
Estimated Shipment: 26-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1085-12-7
Molecular Formula C14 H20 O3
Molecular Weight (g/mol) 236.31
Synonym Benzoic acid, 4-hydroxy-, heptyl ester (9CI, ACI), Benzoic acid, p-hydroxy-, heptyl ester (6CI, 7CI, 8CI), Heptyl alcohol, p-hydroxybenzoate (6CI), Heptyl 4-hydroxybenzoate, Heptyl p-hydroxybenzoate, Heptyl paraben, n-Heptyl 4-hydroxybenzoate, n-Heptyl p-hydroxybenzoate, Nipaheptyl, NSC 309818, Staypro WS 7, 4-Hydroxybenzoic acid heptyl ester
IUPAC Name heptyl 4-hydroxybenzoate
SMILES CCCCCCCOC(=O)c1ccc(O)cc1
Specifications
Quantity 100 g
InChI Formula InChI=1S/C14H20O3/c1-2-3-4-5-6-11-17-14(16)12-7-9-13(15)10-8-12/h7-10,15H,2-6,11H2,1H3
Formula Weight 236.1412
Percent Purity >95%
Chemical Name or Material 4-Hydroxybenzoic acid-n-heptyl ester
Grade HPLC