Learn More
n-Heptadecane-d36, CDN
Specifications
Specifications
| Chemical Name or Material | n-Heptadecane-d36 |
| CAS | 39756-35-9 |
| Molecular Formula | C17 D36 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C17H36/c1-3-5-7-9-11-13-15-17-16-14-12-10-8-6-4-2/h3-17H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,17-hexatriacontadeuterioheptadecane |
| Molecular Weight (g/mol) | 276.69 |
| Formula Weight | 276.508 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |