Learn More
n-Ethyldiisopropylamine, Sigma-Aldrich™
Product Code. 15856270
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856270 1 L 1 L
15846270 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15856270 Supplier Sigma Aldrich Supplier No. 8008941000
 Restricted Item
Estimated Shipment: 24-11-2026
For synthesis
N,N-Diisopropylethylamine (DIPEA), also known as Hunig′s base, is a sterically hindered amine. It is a non-nucleophilic base commonly employed in substitution reactions, alkylations, amide couplings, etc. DIPEA is also used as a base in the Pd-catalyzed cross-coupling reactions, which include Heck coupling and Sonagashira coupling reactions.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7087-68-5
Specifications
Density 0.76 g/cm3 at 20°C
Boiling Point 127°C/1013 hPa
Molecular Formula [(CH3)2CH]2NCH2CH3
MDL Number MFCD00008868
Quantity 1 L
Synonym N-Ethyldiisopropylamine, N,N-Diisopropylethylamine, N-Ethyldiisopropylamine
Molecular Weight (g/mol) 129.24
Grade Research