Learn More
N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide Phosphate Diester, TRC
Product Code. 30384876
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Product Code. Quantity
30384876 100 mg
30376301 250 mg
30378287 25 mg
3 options
Product Code. 30384876
Brand: Toronto Research Chemicals TRCE678835100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2965-52-8
Molecular Formula C24 H19 F34 N2 O8 P S2
Molecular Weight (g/mol) 1204.47
Synonym 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester, diSAmPAP, 1-Octanesulfonamide, N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-N-(2-hydroxyethyl)-, phosphate (ester) (2:1) (8CI), N,N'-[Phosphinicobis(oxy-2,1-ethanediyl)]bis[N-ethyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluoro-1-octanesulfonamide], N-Ethyl-N-(2-hydroxyethyl)perfluorooctanesulfonamide phosphate diester
IUPAC Name bis[2-[ethyl(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylsulfonyl)amino]ethyl] hydrogen phosphate
SMILES CCN(CCOP(=O)(O)OCCN(CC)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C24H19F34N2O8PS2/c1-3-59(70(63,64)23(55,56)19(45,46)15(37,38)11(29,30)9(25,26)13(33,34)17(41,42)21(49,50)51)5-7-67-69(61,62)68-8-6-60(4-2)71(65,66)24(57,58)20(47,48)16(39,40)12(31,32)10(27,28)14(35,36)18(43,44)22(52,53)54/h3-8H2,1-2H3,(H,61,62)
Formula Weight 1203.9778
Chemical Name or Material Bis(2-perfluorooctylsulfonyl-N-ethylaminoethyl) phosphate