missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-2-methyl-2-propen-1-amine, TRC

Product Code. 18030450
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030450 50 mg 50mg
18042250 250 mg 250mg
18001950 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18030450 Supplier Toronto Research Chemicals Supplier No. TRCE92507850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18328-90-0
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.17
Synonym N-Ethyl-2-methyl-allylamine, N-2-Methylallyl-N-ethylamine, N-Ethyl(2-methyl-2-propenyl)amine, N-Ethyl-2-methylallylamine, N-Ethylmethallylamine
SMILES CCNCC(=C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H13N/c1-4-7-5-6(2)3/h7H,2,4-5H2,1,3H3
Formula Weight 99.1048
Chemical Name or Material N-Ethyl-2-methyl-2-propen-1-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.