Learn More
N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride, 95%
Product Code. 11332247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332247 5 g 5g
11322247 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11332247 Supplier Thermo Scientific Alfa Aesar Supplier No. L09744.06
 Restricted Item
In Stock
Add to basket
CAS: 27894-50-4 | C15H23ClN2O4 | 330.81 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27894-50-4
Molecular Formula C15H23ClN2O4
Molecular Weight (g/mol) 330.81
MDL Number MFCD00034846
InChI Key QPNJISLOYQGQTI-ZOWNYOTGSA-N
Synonym h-lys z-ome.hcl, h-lys z-ome hcl, s-methyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-ome hydrochloride, n epsilon-benzyloxycarbonyl-l-lysine methyl ester hydrochloride, nepsilon-cbz-l-lysine methyl ester hydrochloride, methyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, lys z-ome.hcl, h-lys z-ome?cl, h-lys cbz-och3.hcl
PubChem CID 12935371
IUPAC Name methyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
Specifications
Melting Point 115°C to 118°C
Beilstein 3578476
Quantity 5 g
Formula Weight 330.81
Optical Rotation +14.5° (c=2 in Water)
Percent Purity 95%
Chemical Name or Material N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride