Learn More
n-Dodecyltrimethylammonium bromide, MilliporeSigma™
Product Code. 15826780
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15826780 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15826780 Supplier Sigma Aldrich Supplier No. 8410220025
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.348
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide
PubChem CID 14249
ChEBI CHEBI:282662
IUPAC Name dodecyl(trimethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Br-]
Specifications
Melting Point 237°C to 245°C
Color White
Odor Odorless
Quantity 25 g
Formula Weight 308.34
Percent Purity ≥97.0%
Physical Form Solid
Chemical Name or Material n-Dodecyltrimethylammonium bromide