missing translation for 'onlineSavingsMsg'
Learn More

N-dodecane, >99%, MP Biomedicals™

Product Code. 11481042 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
Unit Size:
1 L
100mL
25mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481042 25 mL 25mL
11491042 100 mL 100mL
11401052 500 mL 500mL
11411052 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11481042 Supplier MP Biomedicals Supplier No. 0219515825
  • ONLINE EXCLUSIVE PRICE
    38.60 EUR
    36.75 EUR / 25mL
    Save 1.85 EUR
    4% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Color Colorless
Density 0.753 at 25°C
Assay Percent Range >99%
Quantity 25 mL
Formula Weight 170.34
Percent Purity 0.995
Physical Form Liquid
Chemical Name or Material N-dodecane
compliance-icons compliance-icons
Product Identifier
  • N-Dodecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N-Dodecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.